missing translation for 'onlineSavingsMsg'
Learn More

n-Octadecyltrichlorosilane, 95%, cont. 5-10% branched isomers

CAS: 112-04-9 | C18H37Cl3Si | 387.929 g/mol

$104.51 - $939.14

Chemical Identifiers

CAS 112-04-9
Molecular Formula C18H37Cl3Si
Molecular Weight (g/mol) 387.929
MDL Number MFCD00000484
InChI Key PYJJCSYBSYXGQQ-UHFFFAOYSA-N
Synonym octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane
PubChem CID 8157
IUPAC Name trichloro(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1573218
View Documents
Thermo Scientific Chemicals
A1573218
50 g
Each for $104.51
Only null left
 
AAA1573230
View Documents
Thermo Scientific Chemicals
A1573230
250 g
Each for $310.70
Only null left
 
AAA157320B
View Documents
Thermo Scientific Chemicals
A157320B
1000 g
Each for $939.14
Only null left
 
Description

Description

n-Octadecyltrichlorosilane is used in semiconductor industry to form self-assembled monolayer thin films on silicon dioxide substrates. Used in molecular electronics, as thin insulating gates in Metal-Insulator-Semiconductor Field-Effect Transistors. It is also used in platinum catalyzed synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octadecyltrichlorosilane is used in semiconductor industry to form self-assembled monolayer thin films on silicon dioxide substrates. Used in molecular electronics, as thin insulating gates in Metal-Insulator-Semiconductor Field-Effect Transistors. It is also used in platinum catalyzed synthesis.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Store in cool dry place in tightly closed container. Store under dry inert gas. Keep away from water/moisture.
Specifications

Chemical Identifiers

112-04-9
387.929
PYJJCSYBSYXGQQ-UHFFFAOYSA-N
8157
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
C18H37Cl3Si
MFCD00000484
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane
trichloro(octadecyl)silane

Specifications

160°C to 162°C (3mmHg)
112-04-9
C18H37Cl3Si
MFCD00000484
1778847
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane
PYJJCSYBSYXGQQ-UHFFFAOYSA-N
trichloro(octadecyl)silane
8157
95%
1.46
0.984
cont. 5 to 10% branched isomers
∼20°C
50 g
CH3(CH2)17SiCl3
UN1800
189°C (372°F)
Decomposes in water.
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
387.929
387.94
Pungent
Moisture sensitive
n-Octadecyltrichlorosilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: OCTADECYLTRICHLOROSILANE

EINECSNumber : 203-930-7

RTECSNumber : VV4680000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only