missing translation for 'onlineSavingsMsg'
Learn More

n-Octadecyltrichlorosilane, 95%, cont. 5-10% branched isomers

CAS: 112-04-9 | C18H37Cl3Si | 387.929 g/mol

Supplier:  Thermo Scientific Chemicals A157320B

 View more versions of this product

Catalog No. AAA157320B


Only null left
Explore available promotions

Description

Description

n-Octadecyltrichlorosilane is used in semiconductor industry to form self-assembled monolayer thin films on silicon dioxide substrates. Used in molecular electronics, as thin insulating gates in Metal-Insulator-Semiconductor Field-Effect Transistors. It is also used in platinum catalyzed synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octadecyltrichlorosilane is used in semiconductor industry to form self-assembled monolayer thin films on silicon dioxide substrates. Used in molecular electronics, as thin insulating gates in Metal-Insulator-Semiconductor Field-Effect Transistors. It is also used in platinum catalyzed synthesis.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Store in cool dry place in tightly closed container. Store under dry inert gas. Keep away from water/moisture.
Specifications

Specifications

160°C to 162°C (3mmHg)
∼20°C
1000 g
C18H37Cl3Si
MFCD00000484
1778847
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane
PYJJCSYBSYXGQQ-UHFFFAOYSA-N
trichloro(octadecyl)silane
8157
95%
1.46
0.984
n-Octadecyltrichlorosilane
112-04-9
cont. 5 to 10% branched isomers
CH3(CH2)17SiCl3
UN1800
189°C (372°F)
Decomposes in water.
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
387.929
387.94
Pungent
Moisture sensitive

Chemical Identifiers

112-04-9
387.929
PYJJCSYBSYXGQQ-UHFFFAOYSA-N
8157
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
C18H37Cl3Si
MFCD00000484
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane
trichloro(octadecyl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: OCTADECYLTRICHLOROSILANE

EINECSNumber : 203-930-7

RTECSNumber : VV4680000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only