missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octadecanophenone, 99%, Thermo Scientific™
$51.19 - $51.19
Chemical Identifiers
| CAS | 6786-36-3 |
|---|---|
| Molecular Formula | C24H40O |
| Molecular Weight (g/mol) | 344.583 |
| MDL Number | MFCD00010700 |
| InChI Key | DBLXXVQTWJFJFI-UHFFFAOYSA-N |
| Synonym | octadecanophenone, stearophenone, n-octadecanophenone, heptadecyl phenyl ketone, 1-octadecanone, 1-phenyl, 1-phenyl-1-octadecanone # |
| PubChem CID | 81244 |
| IUPAC Name | 1-phenyloctadecan-1-one |
| SMILES | CCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5339403
|
Thermo Scientific Chemicals
H5339403 |
1 g |
Each for $51.19
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 6786-36-3 | |
| 344.583 | |
| DBLXXVQTWJFJFI-UHFFFAOYSA-N | |
| 81244 | |
| CCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1 |
| C24H40O | |
| MFCD00010700 | |
| octadecanophenone, stearophenone, n-octadecanophenone, heptadecyl phenyl ketone, 1-octadecanone, 1-phenyl, 1-phenyl-1-octadecanone # | |
| 1-phenyloctadecan-1-one |
Specifications
| 6786-36-3 | |
| 425°C | |
| MFCD00010700 | |
| octadecanophenone, stearophenone, n-octadecanophenone, heptadecyl phenyl ketone, 1-octadecanone, 1-phenyl, 1-phenyl-1-octadecanone # | |
| CCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1 | |
| 344.583 | |
| 344.58 | |
| n-Octadecanophenone |
| 65°C to 67°C | |
| C24H40O | |
| 1 g | |
| DBLXXVQTWJFJFI-UHFFFAOYSA-N | |
| 1-phenyloctadecan-1-one | |
| 81244 | |
| 99% |
Safety and Handling
EINECSNumber : 229-850-2
TSCA : No
Recommended Storage : Ambient temperatures