missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl) acetamide, BSA, EMD-Millipore

$568.30 - $568.30

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym BSA
PubChem CID 6913588
IUPAC Name trimethylsilyl N-(trimethylsilyl)ethanimidate
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1096490025
View Documents
Technidata
1096490025
25 mL Glass
Each for $568.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10416-59-8
203.43
SIOVKLKJSOKLIF-UHFFFAOYSA-N
6913588
CC(O[Si](C)(C)C)=N[Si](C)(C)C
C8H21NOSi2
MFCD00008270
BSA
trimethylsilyl N-(trimethylsilyl)ethanimidate

Specifications

Glass
10 hPa (50°C)
C8H21NOSi2
MFCD00008270
SIOVKLKJSOKLIF-UHFFFAOYSA-N
trimethylsilyl N-(trimethylsilyl)ethanimidate
25 mL
203.43
10416-59-8
Density: 0.83g/cm3 (20°C)
CH3C[NSi(CH3)3]OSi(CH3)3
BSA
CC(O[Si](C)(C)C)=N[Si](C)(C)C
203.43
6913588
N, O-Bis(trimethylsilyl) acetamide
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 15° to 25°C

SDS
missing translation for 'documents'

missing translation for 'documents'