missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,O-Bis(trimethylsilyl)acetamide, ≥98.5% (GC), MilliporeSigma™ Supelco™
N,O-Bis(trimethylsilyl)acetamide is an excellent silylation reagent. For some sterically hindered primary or secondary nitro compounds, N,O-bis(trimethylsilyl)acetamide and BSTFA achieve better results than normal silylation conditions.
Supplier: Merck Emd Millipore 1526910X1ML
Specifications
| 71°C to 73°C (35 mmHg) | |
| 10 x 1 mL | |
| MFCD00008270 | |
| BSA | |
| ≥98.5% (GC) | |
| 0.832 g/mL (at 20°C) |
| 10416-59-8 | |
| CH3 C[=NSi(CH3)3 ]OSi(CH3)3 | |
| 44°C | |
| 203.43 | |
| GC Derivatization |
Chemical Identifiers
| MFCD00008270 | |
| BSA |
Safety and Handling
RTECSNumber : AK3000000