missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)acetamide, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187943
Change view
Click to view available options
Quantity:
5 mL
25 mL
10 x 1 mL
Catalog No. Quantity
111020278 5 mL
111018849 10 x 1 mL
111018607 25 mL
3 options

Catalog No. 111020278

Supplier: Merck Emd Millipore 152695ML

Only null left

N,O-Bis(trimethylsilyl)acetamide is an excellent silylation reagent. For some sterically hindered primary or secondary nitro compounds, N,O-bis(trimethylsilyl)acetamide and BSTFA achieve better results than normal silylation conditions.

Chemical Identifiers

MDL Number MFCD00008270
Synonym BSA

Specifications

Boiling Point 71°C to 73°C (35 mmHg)
CAS 10416-59-8
Quantity 5 mL
Linear Formula CH3 C[=NSi(CH3)3 ]OSi(CH3)3
Flash Point 44°C
Formula Weight 203.43
Percent Purity ≥98.5% (GC)
Grade GC Derivatization
Density 0.832 g/mL (at 20°C)
RTECSNumber AK3000000