missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(tert-butyldimethylsilyl)acetamide 96.0+%, TCI America™

Catalog No. B19061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19061G 1 g
B19065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19061G Supplier TCI America Supplier No. B19061G

Chemical Identifiers

CAS 82112-21-8
Molecular Formula C14H33NOSi2
Molecular Weight (g/mol) 287.594
MDL Number MFCD01321138
InChI Key XFPQBAQQJIQJLY-UHFFFAOYSA-N
PubChem CID 529191
IUPAC Name [tert-butyl(dimethyl)silyl] N-[tert-butyl(dimethyl)silyl]ethanimidate
SMILES CC(=N[Si](C)(C)C(C)(C)C)O[Si](C)(C)C(C)(C)C

Specifications

Color Yellow
Quantity 1 g
UN Number 1760
Formula Weight 287.59
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N,O-Bis(tert-butyldimethylsilyl)acetamide [tert-Butyldimethylsilylating Agent]