accessibility menu, dialog, popup

UserName

CH3NSiH3CCH3H3CH3CCH3OSiCH3CH3CH3CH3CH3(E/Z)

CAS RN 82112-21-8

IUPAC Name: tert-butyldimethylsilyl N-(tert-butyldimethylsilyl)ethanimidate
Synonyms: tert-butyldimethylsilyl N-(tert-butyldimethylsilyl)ethanimidate
Molecular Weight (g/mol): 287.59
Molecular Formula: C14H33NOSi2
InChi Key: XFPQBAQQJIQJLY-UHFFFAOYSA-N
SMILES: CC(O[Si](C)(C)C(C)(C)C)=N[Si](C)(C)C(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results