Learn More
n-Nonanoic acid, 97%
CAS: 112-05-0 | C9H18O2 | 158.24 g/mol
$51.82 - $135.37
Chemical Identifiers
| CAS | 112-05-0 |
|---|---|
| Molecular Formula | C9H18O2 |
| Molecular Weight (g/mol) | 158.24 |
| MDL Number | MFCD00004433 |
| InChI Key | FBUKVWPVBMHYJY-UHFFFAOYSA-N |
| Synonym | pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a |
| PubChem CID | 8158 |
| ChEBI | CHEBI:29019 |
| IUPAC Name | nonanoic acid |
| SMILES | CCCCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC356241000
|
N/A | 100 g | Glass bottle |
Each for $51.82
|
|
||||
|
AC356245000
|
N/A | 500 g | Glass bottle |
Each for $72.63
|
|
||||
|
AC356240010
|
N/A | 1 kg | Glass bottle |
Each for $135.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C9H18O2 | |
| MFCD00004433 | |
| pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a | |
| CHEBI:29019 | |
| CCCCCCCCC(=O)O |
Specifications
| 112-05-0 | |
| 100.0 | |
| Colorless | |
| 254.0°C | |
| Authentic | |
| C9H18O2 | |
| CH3(CH2)7CO2H | |
| 02, 352 | |
| 15, 7179 | |
| Solubility in water: negligible. Other solubilities: soluble in alcohol, chloroform and ether, soluble in most other commmon organic solvents | |
| CCCCCCCCC(=O)O | |
| 158.24 | |
| CHEBI:29019 | |
| 97% | |
| n-Nonanoic acid |
| 96.0 | |
| 12.5°C | |
| 0.9000g/mL | |
| 114°C | |
| 96% min. (GC) | |
| 1.4299 to 1.4339 | |
| MFCD00004433 | |
| 0.9 | |
| pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a | |
| FBUKVWPVBMHYJY-UHFFFAOYSA-N | |
| nonanoic acid | |
| 8158 | |
| 158.24 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
EINECSNumber : 203-931-2
RUO – Research Use Only