missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosomorpholine 99.0+%, TCI America™

Catalog No. p-7133825
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04665G 5 g
N046625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04665G Supplier TCI America Supplier No. N04665G
Only null left

Chemical Identifiers

CAS 59-89-2
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00039710
InChI Key ZKXDGKXYMTYWTB-UHFFFAOYSA-N
Synonym n-nitrosomorpholine, nitrosomorpholine, morpholine, 4-nitroso, n-nitrosomorpholin, nmor, morpholine, n-nitroso, n-nitrosomorfolin, n-nitrosomorfolin czech, n-nitrosomorpholin german, unii-3l25fo7fn7
PubChem CID 6046
ChEBI CHEBI:76326
IUPAC Name 4-nitrosomorpholine
SMILES C1COCCN1N=O

Specifications

Color Yellow
Boiling Point 225°C
Quantity 5 g
UN Number 2811
Formula Weight 116.12
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material N-Nitrosomorpholine
RTECSNumber QE7525000
TSCA No