missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosoheptamethyleneimine 99.0+%, TCI America™
Supplier: TCI America N04461G
Specifications
| N-Nitrosoheptamethyleneimine | |
| Yellow | |
| MFCD00059811 | |
| 2811 | |
| RCFKQVLHWYOSFF-UHFFFAOYSA-N | |
| 1-nitrosoazocane | |
| 30360 | |
| ≥99.0% (GC) |
| 20917-49-1 | |
| C7H14N2O | |
| 1 g | |
| Heptamethylenenitrosamine | |
| O=NN1CCCCCCC1 | |
| 142.20 | |
| 142.20 | |
| Crystalline Lumps |
Chemical Identifiers
| 20917-49-1 | |
| 142.20 | |
| RCFKQVLHWYOSFF-UHFFFAOYSA-N | |
| 30360 | |
| O=NN1CCCCCCC1 |
| C7H14N2O | |
| MFCD00059811 | |
| Heptamethylenenitrosamine | |
| 1-nitrosoazocane |
Safety and Handling
RTECSNumber : CN4900000
TSCA : No