missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosoheptamethyleneimine 99.0+%, TCI America™

Catalog No. N0446100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0446100MG 100 mg
N04461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0446100MG Supplier TCI America Supplier No. N0446100MG

Chemical Identifiers

CAS 20917-49-1
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.20
MDL Number MFCD00059811
InChI Key RCFKQVLHWYOSFF-UHFFFAOYSA-N
Synonym Heptamethylenenitrosamine
PubChem CID 30360
IUPAC Name 1-nitrosoazocane
SMILES O=NN1CCCCCCC1

Specifications

Color Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 142.20
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material N-Nitrosoheptamethyleneimine
RTECSNumber CN4900000
TSCA No