missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodipropylamine 98.0+%, TCI America™

Catalog No. p-7133811
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04445G 5 g
N044425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04445G Supplier TCI America Supplier No. N04445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 621-64-7
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.191
MDL Number MFCD00013891
InChI Key YLKFDHTUAUWZPQ-UHFFFAOYSA-N
Synonym n-nitrosodipropylamine, n-nitrosodi-n-propylamine, dipropylnitrosamine, n,n-dipropylnitrosamine, 1-propanamine, n-nitroso-n-propyl, nitrosodipropylamine, dipropylamine, n-nitroso, di-n-propylnitrosamine, di-n-propylnitrosoamine, n-nitroso-n-propyl-1-propanamine
PubChem CID 12130
ChEBI CHEBI:82358
IUPAC Name N,N-dipropylnitrous amide
SMILES CCCN(CCC)N=O

Specifications

Color Yellow
Boiling Point 113°C
Quantity 5 g
UN Number 3082
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Nitrosodipropylamine
RTECSNumber JL9700000
TSCA Yes