missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiphenylamine 98.0+%, TCI America™

$52.19 - $413.83

Chemical Identifiers

CAS 86-30-6
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00019920
InChI Key UBUCNCOMADRQHX-UHFFFAOYSA-N
Synonym n-nitrosodiphenylamine, diphenylnitrosamine, nitrosodiphenylamine, vultrol, benzenamine, n-nitroso-n-phenyl, redax, vulcatard a, retarder j, n-nitroso-n-phenylaniline, ortard
PubChem CID 6838
ChEBI CHEBI:34875
IUPAC Name N-nitroso-N-phenylaniline
SMILES O=NN(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D089925G
View Documents
TCI America
D089925G
25 g N/A
Only null left
 
D0899500G
View Documents
TCI America
D0899500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

86-30-6
198.23
UBUCNCOMADRQHX-UHFFFAOYSA-N
6838
N-nitroso-N-phenylaniline
C12H10N2O
MFCD00019920
n-nitrosodiphenylamine, diphenylnitrosamine, nitrosodiphenylamine, vultrol, benzenamine, n-nitroso-n-phenyl, redax, vulcatard a, retarder j, n-nitroso-n-phenylaniline, ortard
CHEBI:34875
O=NN(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

86-30-6
Green-Yellow
MFCD00019920
3077
UBUCNCOMADRQHX-UHFFFAOYSA-N
N-nitroso-N-phenylaniline
6838
198.23
Crystalline Powder
68°C
C12H10N2O
25 g
n-nitrosodiphenylamine, diphenylnitrosamine, nitrosodiphenylamine, vultrol, benzenamine, n-nitroso-n-phenyl, redax, vulcatard a, retarder j, n-nitroso-n-phenylaniline, ortard
O=NN(C1=CC=CC=C1)C1=CC=CC=C1
198.23
CHEBI:34875
≥98.0% (HPLC)
N-Nitrosodiphenylamine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0431

RTECSNumber : JJ9800000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'