missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosodiphenylamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 68°C |
| Color | Green-Yellow |
| Quantity | 500 g |
| UN Number | 3077 |
| Formula Weight | 198.23 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Nitrosodiphenylamine |
Safety and Handling
| EINECSNumber | (3)-0431 |
| RTECSNumber | JJ9800000 |
| TSCA | Yes |