missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosodiethylamine 99.0+%, TCI America™
Supplier: TCI America D051625ML
Specifications
| N-Nitrosodiethylamine | |
| Yellow | |
| C4H10N2O | |
| 25 mL | |
| n-nitrosodiethylamine, diethylnitrosamine, diethylnitrosoamine, ndea, n-ethyl-n-nitrosoethanamine, n,n-diethylnitrosoamine, nitrosodiethylamine, n,n-diethylnitrosamine, ethanamine, n-ethyl-n-nitroso, diethylnitrosamide | |
| CCN(CC)N=O | |
| 102.14 | |
| CHEBI:34873 | |
| ≥99.0% (GC) |
| 55-18-5 | |
| 177°C | |
| MFCD00013890 | |
| 2810 | |
| WBNQDOYYEUMPFS-UHFFFAOYSA-N | |
| diethyl(nitroso)amine | |
| 5921 | |
| 102.14 | |
| Liquid |
Chemical Identifiers
| 55-18-5 | |
| 102.14 | |
| WBNQDOYYEUMPFS-UHFFFAOYSA-N | |
| 5921 | |
| diethyl(nitroso)amine |
| C4H10N2O | |
| MFCD00013890 | |
| n-nitrosodiethylamine, diethylnitrosamine, diethylnitrosoamine, ndea, n-ethyl-n-nitrosoethanamine, n,n-diethylnitrosoamine, nitrosodiethylamine, n,n-diethylnitrosamine, ethanamine, n-ethyl-n-nitroso, diethylnitrosamide | |
| CHEBI:34873 | |
| CCN(CC)N=O |
Safety and Handling
RTECSNumber : IA3500000
TSCA : Yes