missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiethylamine 99.0+%, TCI America™

Catalog No. p-7126261
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05165ML 5 mL
D051625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05165ML Supplier TCI America Supplier No. D05165ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55-18-5
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
MDL Number MFCD00013890
InChI Key WBNQDOYYEUMPFS-UHFFFAOYSA-N
Synonym n-nitrosodiethylamine, diethylnitrosamine, diethylnitrosoamine, ndea, n-ethyl-n-nitrosoethanamine, n,n-diethylnitrosoamine, nitrosodiethylamine, n,n-diethylnitrosamine, ethanamine, n-ethyl-n-nitroso, diethylnitrosamide
PubChem CID 5921
ChEBI CHEBI:34873
IUPAC Name diethyl(nitroso)amine
SMILES CCN(CC)N=O

Specifications

Color Yellow
Boiling Point 177°C
Quantity 5 mL
UN Number 2810
Formula Weight 102.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Nitrosodiethylamine
RTECSNumber IA3500000
TSCA Yes