missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiethanolamine 97.0+%, TCI America™

$554.44 - $1378.43

Chemical Identifiers

CAS 1116-54-7
Molecular Formula C4H10N2O3
Molecular Weight (g/mol) 134.135
MDL Number MFCD00015522
InChI Key YFCDLVPYFMHRQZ-UHFFFAOYSA-N
Synonym Diethanolnitrosamine, NDELA
PubChem CID 14223
ChEBI CHEBI:82357
IUPAC Name N,N-bis(2-hydroxyethyl)nitrous amide
SMILES C(CO)N(CCO)N=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N04385G
View Documents
TCI America
N04385G
5 g
Each for $554.44
Only null left
 
N043825G
View Documents
TCI America
N043825G
25 g
Each for $1,378.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1116-54-7
134.135
YFCDLVPYFMHRQZ-UHFFFAOYSA-N
14223
N,N-bis(2-hydroxyethyl)nitrous amide
C4H10N2O3
MFCD00015522
Diethanolnitrosamine, NDELA
CHEBI:82357
C(CO)N(CCO)N=O

Specifications

1116-54-7
C4H10N2O3
5 g
YFCDLVPYFMHRQZ-UHFFFAOYSA-N
N,N-bis(2-hydroxyethyl)nitrous amide
14223
134.14
Liquid
114°C
MFCD00015522
Diethanolnitrosamine, NDELA
C(CO)N(CCO)N=O
134.135
CHEBI:82357
≥97.0% (GC)
N-Nitrosodiethanolamine
Videos
Safety and Handling

Safety and Handling

RTECSNumber : KL9550000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'