accessibility menu, dialog, popup

UserName

CAS RN 1116-54-7

IUPAC Name: 2-[(2-hydroxyethyl)(nitroso)amino]ethan-1-ol
Synonyms: N-nitrosodiethanolamine
Molecular Weight (g/mol): 134.14
Molecular Formula: C4H10N2O3
InChi Key: YFCDLVPYFMHRQZ-UHFFFAOYSA-N
SMILES: OCCN(CCO)N=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results