missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodibutylamine 97.0+%, TCI America™

Catalog No. p-7133782
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03755ML 5 mL
N037525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03755ML Supplier TCI America Supplier No. N03755ML

Chemical Identifiers

CAS 924-16-3
Molecular Formula C8H18N2O
Molecular Weight (g/mol) 158.245
MDL Number MFCD00013892
InChI Key YGJHZCLPZAZIHH-UHFFFAOYSA-N
Synonym n-nitrosodibutylamine, dibutylnitrosamine, nitrosodibutylamine, n-nitroso-di-n-butylamine, n-nitrosodi-n-butylamine, 1-butanamine, n-butyl-n-nitroso, ndba, dibutylnitrosoamine, n,n-dibutylnitrosoamine, dbna
PubChem CID 13542
ChEBI CHEBI:82356
IUPAC Name N,N-dibutylnitrous amide
SMILES CCCCN(CCCC)N=O

Specifications

Boiling Point 237°C
Quantity 5 mL
Formula Weight 158.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Nitrosodibutylamine
RTECSNumber EJ4025000
TSCA Yes