missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosoanatabine, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018201
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018201 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018201 Supplier Merck Emd Millipore Supplier No. 752815MG

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706031

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 887407-16-1
Physical Form Neat
Linear Formula C10H11N3 O
MDL Number MFCD01632691
Type Analytical Standard
Quantity 5 mg
Formula Weight 189.21
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard