missing translation for 'onlineSavingsMsg'
Learn More

N-Nicotinoylglycine 98.0+%, TCI America™

Catalog No. p-7133613
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N00925G 5 g
N009225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N00925G Supplier TCI America Supplier No. N00925G
Only null left

Chemical Identifiers

CAS 583-08-4
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00023578
InChI Key ZBSGKPYXQINNGF-UHFFFAOYSA-N
Synonym nicotinuric acid, nicotinylglycine, nicotinoylglycine, 2-nicotinamido acetic acid, n-nicotinoylglycine, n-pyridin-3-ylcarbonyl glycine, nicotinoyl-glycine, n-nicotinylglycine, nicotinurate, n-nicotinoyl-glycine
PubChem CID 68499
ChEBI CHEBI:7563
IUPAC Name 2-(pyridine-3-carbonylamino)acetic acid
SMILES C1=CC(=CN=C1)C(=O)NCC(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 180.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Nicotinoylglycine