missing translation for 'onlineSavingsMsg'
Learn More

N-n-Octylformamide 98.0+%, TCI America™

Catalog No. O03795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03795G 5 g
O037925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03795G Supplier TCI America Supplier No. O03795G

Chemical Identifiers

CAS 6282-06-0
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.26
MDL Number MFCD06252343
InChI Key ZBWPKQRQZDZVSF-UHFFFAOYSA-N
PubChem CID 221394
IUPAC Name N-octylformamide
SMILES CCCCCCCCNC=O

Specifications

Color Yellow
Boiling Point 291°C
Quantity 5 g
Formula Weight 157.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-n-Octylformamide