accessibility menu, dialog, popup

UserName

H3CHNO

CAS RN 6282-06-0

IUPAC Name: N-octylformamide
Synonyms: N-octylformamide
Molecular Weight (g/mol): 157.26
Molecular Formula: C9H19NO
InChi Key: ZBWPKQRQZDZVSF-UHFFFAOYSA-N
SMILES: CCCCCCCCNC=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results