missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-n-Octyl-D-glucamine 98.0+%, TCI America™
Supplier: TCI America O035825G
Specifications
| N-n-Octyl-D-glucamine | |
| 137°C | |
| C14H31NO5 | |
| 25 g | |
| ZRRNJJURLBXWLL-REWJHTLYSA-N | |
| (2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol | |
| 90064 | |
| ≥98.0% (HPLC,T) |
| 23323-37-7 | |
| White-Yellow | |
| MFCD00134352 | |
| n-octyl-d-glucamine, n-octylglucamine, 1-deoxy-1-octylamino-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentaol, n-n-octyl-d-glucamine, 1-octylamino-1-deoxy-d-glucitol, 1-deoxy-1-n-octylamino-d-glucitol, d-glucitol, 1-deoxy-1-octylamino, 1-deoxy-1-octylamine-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentol | |
| CCCCCCCCNCC(C(C(C(CO)O)O)O)O | |
| 293.404 | |
| 293.40 | |
| Crystalline Powder |
Chemical Identifiers
| 23323-37-7 | |
| 293.404 | |
| ZRRNJJURLBXWLL-REWJHTLYSA-N | |
| 90064 | |
| CCCCCCCCNCC(C(C(C(CO)O)O)O)O |
| C14H31NO5 | |
| MFCD00134352 | |
| n-octyl-d-glucamine, n-octylglucamine, 1-deoxy-1-octylamino-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentaol, n-n-octyl-d-glucamine, 1-octylamino-1-deoxy-d-glucitol, 1-deoxy-1-n-octylamino-d-glucitol, d-glucitol, 1-deoxy-1-octylamino, 1-deoxy-1-octylamine-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentol | |
| (2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol |
Safety and Handling
TSCA : No