missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N'-Triphenylbenzidine 97.0+%, TCI America™
Supplier: TCI America T28911G
Specifications
| N,N,N′-Triphenylbenzidine | |
| 167°C | |
| C30H24N2 | |
| 1 g | |
| XHPBZHOZZVRDHL-UHFFFAOYSA-N | |
| N-phenyl-4-[4-(N-phenylanilino)phenyl]aniline | |
| 22013295 | |
| ≥97.0% (GC) |
| 167218-30-6 | |
| Yellow | |
| MFCD17926476 | |
| 4-Diphenylamino-4′C-phenylaminobiphenyl | |
| C1=CC=C(C=C1)NC2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC=C5 | |
| 412.536 | |
| 412.54 | |
| Crystalline Powder |
Chemical Identifiers
| 167218-30-6 | |
| 412.536 | |
| XHPBZHOZZVRDHL-UHFFFAOYSA-N | |
| N-phenyl-4-[4-(N-phenylanilino)phenyl]aniline |
| C30H24N2 | |
| MFCD17926476 | |
| 22013295 | |
| C1=CC=C(C=C1)NC2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC=C5 |
Safety and Handling
TSCA : No