missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N',N″-Triphenyl-1,3,5-benzenetriamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 198°C |
| Quantity | 25 g |
| Formula Weight | 351.45 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N,N′,N′′-Triphenyl-1,3,5-benzenetriamine |
Safety and Handling
| TSCA | No |