accessibility menu, dialog, popup

UserName

CAS RN 102664-66-4

IUPAC Name: N1,N3,N5-triphenylbenzene-1,3,5-triamine
Synonyms: N1,N3,N5-triphenylbenzene-1,3,5-triamine
Molecular Weight (g/mol): 351.45
Molecular Formula: C24H21N3
InChi Key: BMQHYGLEATWRFO-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC(NC2=CC=CC=C2)=CC(NC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results