missing translation for 'onlineSavingsMsg'
Learn More

N,N',N″-Triphenyl-1,3,5-benzenetriamine 98.0+%, TCI America™

Catalog No. p-7136981
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18315G 5 g
T183125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18315G Supplier TCI America Supplier No. T18315G

Chemical Identifiers

CAS 102664-66-4
Molecular Formula C24H21N3
Molecular Weight (g/mol) 351.45
MDL Number MFCD01463903
InChI Key BMQHYGLEATWRFO-UHFFFAOYSA-N
Synonym 1,3,5-Tris(phenylamino)benzene
PubChem CID 227820
IUPAC Name N1,N3,N5-triphenylbenzene-1,3,5-triamine
SMILES N(C1=CC=CC=C1)C1=CC(NC2=CC=CC=C2)=CC(NC2=CC=CC=C2)=C1

Specifications

Melting Point 198°C
Quantity 5 g
Formula Weight 351.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′,N′′-Triphenyl-1,3,5-benzenetriamine