missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N'-Trimethylethylenediamine 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 116°C |
| Quantity | 5 mL |
| UN Number | 2733 |
| Formula Weight | 102.18 |
| Percent Purity | ≥97.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | N,N,N′-Trimethylethylenediamine |
Safety and Handling
| TSCA | Yes |