accessibility menu, dialog, popup

UserName

H3CH2NNHCH3CH3

CAS RN 142-25-6

IUPAC Name: [2-(dimethylazaniumyl)ethyl](methyl)azanium
Synonyms: [2-(dimethylammonio)ethyl](methyl)azanium
Molecular Weight (g/mol): 104.2
Molecular Formula: C5H16N2
InChi Key: HVOYZOQVDYHUPF-UHFFFAOYSA-P
SMILES: C[NH2+]CC[NH+](C)C
Boiling Point 
  • (2)
  • (6)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (2)