missing translation for 'onlineSavingsMsg'
Learn More

N,N',N″-Triacetylchitotriose 98.0+%, TCI America™

Catalog No. T291220MG
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T291220MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T291220MG Supplier TCI America Supplier No. T291220MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38864-21-0
Molecular Formula C24H41N3O16
Molecular Weight (g/mol) 627.597
MDL Number MFCD00136047
InChI Key LRDDKCYRFNJZBX-UHFFFAOYSA-N
Synonym Tri-N-acetylchitotriose, GlcNAc beta(1-4)GlcNAc beta(1-4)GlcNAc
PubChem CID 5070942
IUPAC Name N-[2-[5-acetamido-6-(5-acetamido-1,2,4-trihydroxy-6-oxohexan-3-yl)oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)NC(=O)C)OC(C(CO)O)C(C(C=O)NC(=O)C)O)CO)CO)O)O

Specifications

Melting Point 311°C
Color White
Quantity 20 mg
Formula Weight 627.60
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′,N′′-Triacetylchitotriose
TSCA No
Recommended Storage Freezer