CAS RN 38864-21-0
IUPAC Name:
N-[(2R,3R,4S,5R)-4-{[(2S,3R,4R,5S,6R)-3-acetamido-5-{[(2S,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3,5,6-trihydroxy-1-oxohexan-2-yl]acetamide
Synonyms:
N-[(2R,3R,4S,5R)-4-{[(2S,3R,4R,5S,6R)-3-acetamido-5-{[(2S,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3,5,6-trihydroxy-1-oxohexan-2-yl]acetamide
Molecular Weight (g/mol):
627.6
Molecular Formula:
C24H41N3O16
InChi Key:
LRDDKCYRFNJZBX-WHFMPQCRSA-N
SMILES:
CC(=O)N[C@@H](C=O)[C@@H](O)[C@H](O[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2NC(C)=O)[C@H](O)[C@H]1NC(C)=O)[C@H](O)CO
1
–
1
of
1
results
1
N,N',N″-Triacetylchitotriose 98.0+%, TCI America™
CAS: 38864-21-0 Molecular Formula: C24H41N3O16 Molecular Weight (g/mol): 627.597 MDL Number: MFCD00136047 InChI Key: LRDDKCYRFNJZBX-UHFFFAOYSA-N Synonym: Tri-N-acetylchitotriose, GlcNAc beta(1-4)GlcNAc beta(1-4)GlcNAc PubChem CID: 5070942 IUPAC Name: N-[2-[5-acetamido-6-(5-acetamido-1,2,4-trihydroxy-6-oxohexan-3-yl)oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide SMILES: CC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)NC(=O)C)OC(C(CO)O)C(C(C=O)NC(=O)C)O)CO)CO)O)O