missing translation for 'onlineSavingsMsg'
Learn More

N,N',N″-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine 98.0+%, TCI America™

Catalog No. t32401g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
Catalog No. Quantity
T32401G 1 g
T3240200MG 200 mg
2 options

Catalog No. T32401G

Supplier: TCI America T32401G

Only null left

Chemical Identifiers

CAS 82504-70-9
Molecular Formula C24H24N6
Molecular Weight (g/mol) 396.498
InChI Key XMOZWTZDMACVMS-UHFFFAOYSA-N
Synonym Tris(3-methylanilino)triazine
PubChem CID 11784255
IUPAC Name 2-N,4-N,6-N-tris(3-methylphenyl)-1,3,5-triazine-2,4,6-triamine
SMILES CC1=CC(=CC=C1)NC2=NC(=NC(=N2)NC3=CC=CC(=C3)C)NC4=CC=CC(=C4)C

Specifications

Melting Point 233°C
Color White-Yellow
Quantity 1 g
Formula Weight 396.50
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′,N′′-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine
EINECSNumber (5)-6755
TSCA No