accessibility menu, dialog, popup

UserName

CAS RN 82504-70-9

IUPAC Name: N2,N4,N6-tris(3-methylphenyl)-1,3,5-triazine-2,4,6-triamine
Synonyms: N2,N4,N6-tris(3-methylphenyl)-1,3,5-triazine-2,4,6-triamine
Molecular Weight (g/mol): 396.5
Molecular Formula: C24H24N6
InChi Key: XMOZWTZDMACVMS-UHFFFAOYSA-N
SMILES: CC1=CC(NC2=NC(NC3=CC=CC(C)=C3)=NC(NC3=CC=CC(C)=C3)=N2)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results