missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraphenyl-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. p-7137690
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33091G 1 g
T33095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33091G Supplier TCI America Supplier No. T33091G
Only null left

Chemical Identifiers

CAS 14118-16-2
Molecular Formula C30H24N2
Molecular Weight (g/mol) 412.536
MDL Number MFCD08276846
InChI Key JPDUPGAVXNALOL-UHFFFAOYSA-N
Synonym 1,4-Bis(diphenylamino)benzene
PubChem CID 84207
IUPAC Name 1-N,1-N,4-N,4-N-tetraphenylbenzene-1,4-diamine
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Melting Point 203°C
Color White-Yellow
Quantity 1 g
Formula Weight 412.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetraphenyl-1,4-phenylenediamine