CAS RN 14118-16-2
CAS RN 14118-16-2
IUPAC Name:
N1,N1,N4,N4-tetraphenylbenzene-1,4-diamine
Synonyms:
N1,N1,N4,N4-tetraphenylbenzene-1,4-diamine
Molecular Weight (g/mol):
412.54
Molecular Formula:
C30H24N2
InChi Key:
JPDUPGAVXNALOL-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
N,N,N',N'-Tetraphenyl-1,4-phenylenediamine 98.0+%, TCI America™
CAS: 14118-16-2 Molecular Formula: C30H24N2 Molecular Weight (g/mol): 412.536 MDL Number: MFCD08276846 InChI Key: JPDUPGAVXNALOL-UHFFFAOYSA-N Synonym: 1,4-Bis(diphenylamino)benzene PubChem CID: 84207 IUPAC Name: 1-N,1-N,4-N,4-N-tetraphenylbenzene-1,4-diamine SMILES: C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC=C5