missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetraphenyl-1,4-phenylenediamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 203°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 412.54 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N,N,N′,N′-Tetraphenyl-1,4-phenylenediamine |
Safety and Handling
| TSCA | Yes |