missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylthiourea, 98%

Catalog No. p-7046183
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1339206 5 g
AAL1339214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1339206 Supplier Thermo Scientific Chemicals Supplier No. L1339206
Only null left

CAS: 2782-91-4 | C5H12N2S | 132.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2782-91-4
Molecular Formula C5H12N2S
Molecular Weight (g/mol) 132.23
MDL Number MFCD00008324
InChI Key MNOILHPDHOHILI-UHFFFAOYSA-N
Synonym tetramethylthiourea, thiourea, tetramethyl, basthioryl, 1,1,3,3-tetramethyl-2-thiourea, tmtu, n,n,n',n'-tetramethylthiourea, urea, thio-, tetramethyl, tetramethyl-2-thiourea, urea, 1,1,3,3-tetramethyl-2-thio, thiourea, n,n,n',n'-tetramethyl
PubChem CID 17725
IUPAC Name 1,1,3,3-tetramethylthiourea
SMILES CN(C)C(=S)N(C)C

Specifications

Melting Point 75°C to 77°C
Boiling Point 245°C
Flash Point 190°C (374°F)
Odor Odorless
Quantity 5 g
Beilstein 1744916
Formula Weight 132.23
Percent Purity 98%
Chemical Name or Material N,N,N',N'-Tetramethylthiourea
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 220-488-0
RTECSNumber YU2750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only