missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylethylenediamine (TEMED), Hoefer™

Catalog No. 03500534
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03500534 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03500534 Supplier Hoefer Inc Supplier No. GR15125

missing translation for 'validMainframeMsg'

Catalyzes the formation of free radicals by ammonium persulfate and accelerates the polymerization of acrylamide and bisacrylamide

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Color Colorless to Yellow
Density 0.77 ± 0.01(20°C)
Assay Percent Range ≥98%
Linear Formula (CH3)2NCH2CH2N(CH3)2
Quantity 25 mL
Formula Weight 116.20
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine