missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethylethylenediamine 98.0+%, TCI America™
Supplier: TCI America T0147100ML
Specifications
| N,N,N′,N′-Tetramethylethylenediamine | |
| -55°C | |
| 122°C | |
| MFCD00008335 | |
| 2372 | |
| KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
| N,N,N',N'-tetramethylethane-1,2-diamine | |
| 8037 | |
| 116.21 | |
| Liquid |
| 110-18-9 | |
| Colorless | |
| C6H16N2 | |
| 100 mL | |
| temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
| CN(C)CCN(C)C | |
| 116.208 | |
| CHEBI:32850 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 110-18-9 | |
| 116.208 | |
| KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
| 8037 | |
| N,N,N',N'-tetramethylethane-1,2-diamine |
| C6H16N2 | |
| MFCD00008335 | |
| temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
| CHEBI:32850 | |
| CN(C)CCN(C)C |
Safety and Handling
EINECSNumber : (2)-0155
RTECSNumber : KV7175000
TSCA : Yes