missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylethylenediamine 98.0+%, TCI America™

Catalog No. p-7137255
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25155G 5 g
T251525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25155G Supplier TCI America Supplier No. T25155G

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Melting Point -55°C
Color Yellow
Boiling Point 122°C
Quantity 5 g
UN Number 2372
Formula Weight 116.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetramethylethylenediamine [for Electrophoresis]
EINECSNumber (2)-0155
RTECSNumber KV7175000
TSCA Yes