Learn More
Thermo Scientific Chemicals N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade
Description
N,N,N',N'-Tetramethylethylenediamine is used as polymerization accelerator in gel electrophoresis, solvent and oxidizing reagent. N,N,N',N'-Tetramethylethylenediamine is used as anti-hyperlipidemic, plant growth stimulator, pathogen growth inhibitor, confers plant disease resistance. TEMED is used with ammonium persulfate to catalyze the polymerization of acrylamide when making polyacrylamide gels, used in gel electrophoresis. TMEDA is widely employed as a ligand for metal ions. TEMED can also be a component of Hypergolic propellants. Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is frequently used with another catalyst, APS, for the preparation of polyacrylamide gels for protein and nucleic acid analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N,N′,N′-Tetramethylethylenediamine is used as polymerization accelerator in gel electrophoresis, solvent and oxidizing reagent. N,N,N′,N′-Tetramethylethylenediamine is used as anti-hyperlipidemic, plant growth stimulator, pathogen growth inhibitor, confers plant disease resistance. TEMED is used with ammonium persulfate to catalyze the polymerization of acrylamide when making polyacrylamide gels, used in gel electrophoresis. TMEDA is widely employed as a ligand for metal ions. TEMED can also be a component of Hypergolic propellants. Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is frequently used with another catalyst, APS, for the preparation of polyacrylamide gels for protein and nucleic acid analysis.
Solubility
Soluble in water 10 mg/mL at 20°C.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, copper, mercury, water, moisture.
Specifications
Specifications
| Melting Point | -55°C |
| Color | Colorless |
| Density | 0.775 |
| Boiling Point | 120°C to 122°C |
| Flash Point | 18°C (154°F) |
| Odor | Unpleasant |
| Refractive Index | 1.418 |
| Linear Formula | (CH3)2NCH2CH2N(CH3)2 |
| Quantity | 25 mL |
| UN Number | UN2372 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H225-H302+H332-H314-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H225-H314-H302-H332 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Harmful if swallowed. Harmful if inhaled. |
| missing translation for 'healthHazard3' | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DI-(DIMETHYLAMINO)ETHANE |
| EINECSNumber | 203-744-6 |
| RTECSNumber | KV7175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only