missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylethylenediamine, 99%

CAS: 110-18-9 | C6H16N2 | 116.208 g/mol

Supplier:  Thermo Scientific Chemicals A125360F

 View more versions of this product

Catalog No. AAA125360F

Only null left
Explore available promotions

Description

Description

N,N,N',N'-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.

Solubility
Miscible with water.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, carbon dioxide and copper.
Specifications

Specifications

N,N,N',N'-Tetramethylethylenediamine
-55°C
120°C to 122°C
99%
C6H16N2
(CH3)2NCH2CH2N(CH3)2
2500 mL
1732991
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
KWYHDKDOAIKMQN-UHFFFAOYSA-N
N,N,N',N'-tetramethylethane-1,2-diamine
8037
116.21
Liquid
110-18-9
0.775
18°C (62°F)
Unpleasant
1.418
MFCD00008335
UN2372
Air sensitive; Hygroscopic
Miscible with water.
CN(C)CCN(C)C
116.208
CHEBI:32850
99%

Chemical Identifiers

110-18-9
116.208
KWYHDKDOAIKMQN-UHFFFAOYSA-N
8037
N,N,N',N'-tetramethylethane-1,2-diamine
C6H16N2
MFCD00008335
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
CHEBI:32850
CN(C)CCN(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H302-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H332-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DI-(DIMETHYLAMINO)ETHANE

EINECSNumber : 203-744-6

RTECSNumber : KV7175000

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only