missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N,N',N'-Tetramethylethylenediamine, 99%, for biochemistry, For electrophoresis

Catalog No. AC420580050
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420580050 5 g Glass bottle
AC420580500 50 g Glass bottle
AC420581000 100 g Glass bottle
AC420585000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC420580050 Supplier Thermo Scientific Chemicals Supplier No. 420580050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-18-9 | C6H16N2 | 116.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -55.0°C
Color Colorless to Yellow
Density 0.7700g/mL
Boiling Point 120.0°C to 122.0°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4169 to 1.4189
Linear Formula (CH3)2NCH2CH2N(CH3)2
Quantity 5 g
Beilstein 04, 250
Fieser 02,403; 03,284; 04,485; 05,652; 06,576; 07,358; 11,340; 12,477; 14,68; 15,63; 16,55
Specific Gravity 0.77
Merck Index 15, 9277
Solubility Information (10% in water) Clear and no haze
Formula Weight 116.21
Percent Purity 99%
Grade Biochemistry
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-744-6
RTECSNumber KV7175000
TSCA TSCA

RUO – Research Use Only