missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethyldiaminomethane 98.0+%, TCI America™
Supplier: TCI America T0969100ML
Specifications
| N,N,N′,N′-Tetramethyldiaminomethane | |
| Colorless | |
| C5H14N2 | |
| 100 mL | |
| n,n,n',n'-tetramethyldiaminomethane, bis dimethylamino methane, methanediamine, n,n,n',n'-tetramethyl, tetramethylmethylenediamine, methylenebis dimethylamine, n,n,n',n'-tetramethylmethylenediamine, unii-z870i525ks, methylenediamine, n,n,n',n'-tetramethyl, dimethylamino methyl dimethylamine, dimethyl dimethylamino methyl amine | |
| CN(C)CN(C)C | |
| 102.18 | |
| 102.18 | |
| Liquid |
| 51-80-9 | |
| 84°C | |
| MFCD00008328 | |
| 2733 | |
| VGIVLIHKENZQHQ-UHFFFAOYSA-N | |
| [(dimethylamino)methyl]dimethylamine | |
| 5829 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 51-80-9 | |
| 102.18 | |
| VGIVLIHKENZQHQ-UHFFFAOYSA-N | |
| 5829 | |
| CN(C)CN(C)C |
| C5H14N2 | |
| MFCD00008328 | |
| n,n,n',n'-tetramethyldiaminomethane, bis dimethylamino methane, methanediamine, n,n,n',n'-tetramethyl, tetramethylmethylenediamine, methylenebis dimethylamine, n,n,n',n'-tetramethylmethylenediamine, unii-z870i525ks, methylenediamine, n,n,n',n'-tetramethyl, dimethylamino methyl dimethylamine, dimethyl dimethylamino methyl amine | |
| [(dimethylamino)methyl]dimethylamine |
Safety and Handling
RTECSNumber : PA6700000
TSCA : Yes