missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyldiaminomethane 98.0+%, TCI America™

Catalog No. p-7136524
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T096925ML 25 mL
T0969100ML 100 mL
T0969500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T096925ML Supplier TCI America Supplier No. T096925ML
Only null left

Chemical Identifiers

CAS 51-80-9
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008328
InChI Key VGIVLIHKENZQHQ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyldiaminomethane, bis dimethylamino methane, methanediamine, n,n,n',n'-tetramethyl, tetramethylmethylenediamine, methylenebis dimethylamine, n,n,n',n'-tetramethylmethylenediamine, unii-z870i525ks, methylenediamine, n,n,n',n'-tetramethyl, dimethylamino methyl dimethylamine, dimethyl dimethylamino methyl amine
PubChem CID 5829
IUPAC Name [(dimethylamino)methyl]dimethylamine
SMILES CN(C)CN(C)C

Specifications

Color Colorless
Boiling Point 84°C
Quantity 25 mL
UN Number 2733
Formula Weight 102.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetramethyldiaminomethane
RTECSNumber PA6700000
TSCA Yes