Learn More
N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 212333-72-7 |
|---|---|
| Molecular Formula | C10H16N3OS·F6P |
| Molecular Weight (g/mol) | 371.29 |
| InChI Key | UCOGEMMJHLHOAW-UHFFFAOYSA-N |
| Synonym | hott, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate, hott n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, ambotzrl-1156, acmc-209fij, ksc911c8f, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuron, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronhexafluorophosphate, n,n,n,n-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate |
| PubChem CID | 10571248 |
| IUPAC Name | [dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate |
| SMILES | CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC440260050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Coupling reagentChemical Identifiers
| 212333-72-7 | |
| 371.29 | |
| hott, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate, hott n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, ambotzrl-1156, acmc-209fij, ksc911c8f, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuron, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronhexafluorophosphate, n,n,n,n-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate | |
| [dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate |
| C10H16N3OS·F6P | |
| UCOGEMMJHLHOAW-UHFFFAOYSA-N | |
| 10571248 | |
| CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F |
Specifications
| 212333-72-7 | |
| 100.0 | |
| Authentic | |
| Glass Bottle | |
| hott, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate, hott n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, ambotzrl-1156, acmc-209fij, ksc911c8f, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuron, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronhexafluorophosphate, n,n,n,n-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate | |
| CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F | |
| 371.29 | |
| 371.29 | |
| N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate |
| 96.0 | |
| 30.41 to 34.29% C, 10.64 to 12.00% N | |
| 97% | |
| C10H16N3OS·F6P | |
| UCOGEMMJHLHOAW-UHFFFAOYSA-N | |
| [dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate | |
| 10571248 | |
| 97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only