accessibility menu, dialog, popup

UserName

H3CNCH3NCH3CH3SNOFPFFFFF

CAS RN 212333-72-7

IUPAC Name: hexafluoro-λ⁵-phosphanuide 2-{[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}pyridin-1-ium-1-olate
Synonyms: 2-{[(dimethylamino)(dimethyliminio)methyl]sulfanyl}pyridin-1-ium-1-olate hexafluorophosphate
Molecular Weight (g/mol): 371.28
Molecular Formula: C10H16F6N3OPS
InChi Key: UCOGEMMJHLHOAW-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CN(C)C(SC1=CC=CC=[N+]1[O-])=[N+](C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)