missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiouronium Tetrafluoroborate 98.0+%, TCI America™
Supplier: TCI America T356925G
Specifications
| N,N,N′,N′-Tetramethyl-S-(1-oxido-2-pyridyl)thiouronium Tetrafluoroborate | |
| 108°C | |
| C10H16BF4N3OS | |
| 25 g | |
| LHLFXDQURZVFLK-UHFFFAOYSA-N | |
| tetrafluoroboranuide 2-{[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}pyridin-1-ium-1-olate | |
| 10543173 | |
| ≥98.0% (HPLC) |
| 255825-38-8 | |
| White-Yellow | |
| MFCD05664718 | |
| tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, 2-dimethylamino dimethyliminio methyl sulfanyl pyridin-1-ium-1-olate tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate, s-2-pyridyl-n,n,n',n'-tetramethylthiuronium n-oxide tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate nmr, tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, dimethylamino-1-oxidopyridin-1-ium-2-yl sulfanyl-methylene-dimethyl-ammonium tetrafluoroborate | |
| F[B-](F)(F)F.CN(C)C(SC1=CC=CC=[N+]1[O-])=[N+](C)C | |
| 313.12 | |
| 313.12 | |
| Crystalline Powder |
Chemical Identifiers
| 255825-38-8 | |
| 313.12 | |
| LHLFXDQURZVFLK-UHFFFAOYSA-N | |
| 10543173 | |
| F[B-](F)(F)F.CN(C)C(SC1=CC=CC=[N+]1[O-])=[N+](C)C |
| C10H16BF4N3OS | |
| MFCD05664718 | |
| tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, 2-dimethylamino dimethyliminio methyl sulfanyl pyridin-1-ium-1-olate tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium tetrafluoroborate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate, s-2-pyridyl-n,n,n',n'-tetramethylthiuronium n-oxide tetrafluoroborate, s-1-oxido-2-pyridyl-n,n,n',n'-tetramethylthiuronium tetrafluoroborate nmr, tott, 2-1-oxy-pyridin-2-yl-1,1,3,3-tetramethylisothiouronium tetrafluoroborate, dimethylamino-1-oxidopyridin-1-ium-2-yl sulfanyl-methylene-dimethyl-ammonium tetrafluoroborate | |
| tetrafluoroboranuide 2-{[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}pyridin-1-ium-1-olate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)